Learn More
Apexbio Technology LLC Alprostadil(Synonyms: Prostaglandin E1, PGE1, Alprostadilum, Edex, Caverject, Muse, Prostin VR, Liprostin), 10mg, CAS: 745-65-3.
SDP
Catalog No. 502239982
Encompass_Preferred
Alprostadil (CAS 745-65-3) also known as prostaglandin E1 is an endogenous prostaglandin analog that exerts potent vasodilatory activity by binding and activating prostaglandin E-type (EP) receptors primarily EP2 and EP4 Upon receptor activation intracellular cyclic adenosine monophosphate (cAMP) levels rise mediating smooth muscle relaxation and vasodilation Clinically Alprostadil is investigated and utilized in research contexts addressing vascular biology erectile dysfunction mechanisms and cardiovascular function offering insights into prostaglandin signaling pathways
This item is not returnable. View return policy
Catalog No. 50-223-9982 Supplier Apexbio Technology LLC Supplier No. B215410
Alprostadil (CAS 745-65-3) also known as prostaglandin E1 is an endogenous prostaglandin analog that exerts potent vasodilatory activity by binding and activating prostaglandin E-type (EP) receptors primarily EP2 and EP4 Upon receptor activation intracellular cyclic adenosine monophosphate (cAMP) levels rise mediating smooth muscle relaxation and vasodilation Clinically Alprostadil is investigated and utilized in research contexts addressing vascular biology erectile dysfunction mechanisms and cardiovascular function offering insights into prostaglandin signaling pathways