Learn More
Medchemexpress LLC Alrestatin | 51411-04-2 | 99.7% | 250 MG
SDP
Catalog No. 5000414332
Encompass_Preferred
Alrestatin is an inhibitor of aldose reductase, an enzyme involved in the pathogenesis of complications of diabetes mellitus, including diabetic neuropathy. It is intended for research use only and not for patient use.
  • Inhibits aldose reductase
  • Intended for research use only
  • Not for patient use
  • White to off-white solid appearance
Catalog No. 50-004-14332 Supplier Medchemexpress LLC Supplier No. 250MGHYB1202
Alrestatin is an inhibitor of aldose reductase, an enzyme involved in the pathogenesis of complications of diabetes mellitus, including diabetic neuropathy. It is intended for research use only and not for patient use.
  • Inhibits aldose reductase
  • Intended for research use only
  • Not for patient use
  • White to off-white solid appearance