Learn More
Medchemexpress LLC HY-101518 5mg , Alrizomadlin CAS:1818393-16-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015185MG
Encompass_Preferred
HY-101518 5mg Alrizomadlin CAS: 1818393-16-6 Alrizomadlin (APG-115) is an orally active MDM2 protein inhibitor binding to MDM2 protein with IC50 and Ki values of 3.8 nM and 1 nM, respectively. Alrizomadlin blocks the interaction of MDM2 and p53 and induces cell-cycle arrest and apoptosis in a p53-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9887