Learn More
Medchemexpress LLC HY-12983 1mg Medchemexpress, ALS-8112 CAS:1445379-92-9 Purity:>98%
SDP
Catalog No. 502081222
Encompass_Preferred
Medchemexpress, HY-12983 1mg ALS-8112 CAS:1445379-92-9 ALS-8112 is a potent and selective respiratory syncytial virus (RSV) polymerase inhibitor. The 5'-triphosphate form of ALS-8112 inhibits RSV polymerase with an IC50 of 0.02 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1222 Supplier Medchemexpress LLC Supplier No. HY129831MG
Medchemexpress, HY-12983 1mg ALS-8112 CAS:1445379-92-9 ALS-8112 is a potent and selective respiratory syncytial virus (RSV) polymerase inhibitor. The 5'-triphosphate form of ALS-8112 inhibits RSV polymerase with an IC50 of 0.02 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.