Learn More
Aviva Systems Biology Corp ALS2 Antibody - middle region (ARP57487_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57487_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ALS2. Alternate names: ALS2CR6, ALSJ, FLJ31851, IAHSP, KIAA1563, MGC87187, PLSJApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54042