Learn More
Aviva Systems Biology Corp C2CD6 Antibody - N-terminal region (ARP53022_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53022_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALS2CR11. It was validated on Western Blot. . Alternate names: DKFZp686H0636, FLJ25351, FLJ40332ApplicationWB. Species reactivity: Human
Catalog No. 50-285-794