Learn More
Electron Microscopy Sciences Anti Vibration Table ALSC ALSC Acoustic Hood, 75 x 75 cm
SDP
Catalog No. 501930092
Encompass_Preferred
Remove the airborne vibrations that limit the performance of sensitive instruments. A unique construction using interlocking acrylic panels gives complete visibility during work operations. Front panel with portholes for hand access, lifts off to allow easy cleaning of acrylic panels and stainless worktops.
Size: 75 x 75 cm
Catalog No. 50-193-0092 Supplier Electron Microscopy Sciences Supplier No. 9962
Remove the airborne vibrations that limit the performance of sensitive instruments. A unique construction using interlocking acrylic panels gives complete visibility during work operations. Front panel with portholes for hand access, lifts off to allow easy cleaning of acrylic panels and stainless worktops.
Size: 75 x 75 cm