Learn More
ILEX LIFE SCIENCES LLC ALSEVERS SOLUTION 1000 ML
SDP
Catalog No. 502733787
Encompass_Preferred
PAN-Biotech Alsever's Solution (1000 ml). Ca/Mg: Without calcium and magnesium. BICARBONATE: Without sodium bicarbonate. PHENOL RED: Without Phenol Red. STERILE: Yes. STORAGE: +2-8°C. PRODUCT CATEGORY: Balanced salt solutions. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: PAN-Biotech Alsever's Solution is an isotonic balanced solution. It is used as an anticoagulant or blood preservative. Alsever's Solution enables whole blood to be stored at refrigerator temperature up to 10 weeks.
Catalog No. 50-273-3787 Supplier ILEX LIFE SCIENCES LLC Supplier No. P04421000
Add to Cart
Add to Cart
PAN-Biotech Alsever's Solution (1000 ml). Ca/Mg: Without calcium and magnesium. BICARBONATE: Without sodium bicarbonate. PHENOL RED: Without Phenol Red. STERILE: Yes. STORAGE: +2-8°C. PRODUCT CATEGORY: Balanced salt solutions. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: PAN-Biotech Alsever's Solution is an isotonic balanced solution. It is used as an anticoagulant or blood preservative. Alsever's Solution enables whole blood to be stored at refrigerator temperature up to 10 weeks.