Learn More
Cayman Chemical Alisol A, Purity: ≥98%, CAS: 19885-10-0, 10 mg
SDP
Catalog No. 502950362
Encompass_Preferred
A terpenoid; inhibits HBV surface antigen secretion in HBV-infected 2.2.15 cells (IC50 = 39 µM); induces cell cycle arrest at the G0/G1 phase and autophagy in MDA-MB-231 human breast cancer cells at 20 and 40 µM; reduces hepatic steatosis and increases in plasma triglyceride, total cholesterol, LDL-C, and HDL-C levels in a mouse model of high-fat diet-induced obesity at 100 mg/kg per day; decreases blood glucose and insulin levels in intraperitoneal glucose and insulin tolerance tests, respectively, in the same model
Catalog No. 50-295-0362 Supplier Cayman Chemical Supplier No. 3405010 MG
A terpenoid; inhibits HBV surface antigen secretion in HBV-infected 2.2.15 cells (IC50 = 39 µM); induces cell cycle arrest at the G0/G1 phase and autophagy in MDA-MB-231 human breast cancer cells at 20 and 40 µM; reduces hepatic steatosis and increases in plasma triglyceride, total cholesterol, LDL-C, and HDL-C levels in a mouse model of high-fat diet-induced obesity at 100 mg/kg per day; decreases blood glucose and insulin levels in intraperitoneal glucose and insulin tolerance tests, respectively, in the same model