Learn More
Cayman Chemical Alsol B 10mg
SDP
Catalog No. 502910983
Encompass_Preferred
Source Active recombinant SARS-CoV-2 N-terminal His-tagged papain-like protease expressed in E coli Amino acids 1564-1880 MW 36 8 kDa
Catalog No. 50-291-0983 Supplier Cayman Chemical Supplier No. 34808 10MG
Source Active recombinant SARS-CoV-2 N-terminal His-tagged papain-like protease expressed in E coli Amino acids 1564-1880 MW 36 8 kDa