Learn More
Medchemexpress LLC HY-108359 10mg Medchemexpress, Alsterpaullone CAS:237430-03-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10835910MG
Encompass_Preferred
Medchemexpress, HY-108359 10mg Alsterpaullone CAS:237430-03-4 Alsterpaullone (9-Nitropaullone;NSC 705701) is a potent CDK inhibitor, with IC50s of 35 nM, 15 nM, 200 nM and 40 nM for CDK1/cyclin B, CDK2/cyclin A, CDK2/cyclin E and CDK5/p35, respectively. Alsterpaullone also competes with ATP for binding to GSK-3alpha/GSK-3beta with IC50s of both 4 nM. Alsterpaullone has antitumor activity, and possesses potential for the treatment of neurodegenerative and proliferative disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8538