Learn More
Enzo Life Sciences Alsterpaullone (5mg). CAS: 237430-03-4
SDP
Supplier:  Enzo Life Sciences ALX270275M005
Encompass_Preferred
Potent inhibitor of CDK1/cyclin B (IC50=35nM). Potent and selective inhibitor of CDK2/cyclin A, CDK2/cyclin E (IC50=200nM), CDK5/p25 (IC50=40nM), CDK5/p35 (IC50=40nM) and GSK-3β . Induces apoptosis by activation of caspase-8 and caspase-9 followed by disruption of the mitochondrial potential.Licensed from NCI. Formula: C16H11N3O3. MW: 293.3. Purity: ≥95%. Solubility: Soluble in DMSO; insoluble in water or 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-8457