Learn More
Mac-Mod Avantor Alltima C18, 100 A, 5 um, 4.6 x 250 mm HPLC Column-W (Waters Port)
SDP
Catalog No. 501120363
Encompass
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.
Catalog No. 50-112-0363 Supplier Mac-Mod Supplier No. 88057
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.