Learn More
Teco Diagnostics ALT/GPT LIQUID(KINETIC)150TEST
Catalog No. NC9851323
Encompass_Preferred
For the quantitative determination of alanine aminotransferase in serum used in routine examination and monitoring of therapy and relapses. 1x125mL Bottle of ALT (SGPT) Liquid Reagent 1, 1x25mL Bottle of ALT (SGPT) Liquid Reagent 2
Catalog No. NC9851323 Supplier Teco Diagnostics Supplier No. A524150
For the quantitative determination of alanine aminotransferase in serum used in routine examination and monitoring of therapy and relapses. 1x125mL Bottle of ALT (SGPT) Liquid Reagent 1, 1x25mL Bottle of ALT (SGPT) Liquid Reagent 2