Learn More
Integrated DNA Technologies Inc Alt-R S.p. Cas9 Nuclease V3, High Purity, RNP Delivery, 100 µg
Supplier:  Integrated DNA Technologies Inc 1081058
Encompass
Alt-R S.p. Cas9 Nuclease V3 is a high-purity, recombinant Cas9 enzyme from *Streptococcus pyogenes*. Engineered for efficient genome editing, it enhances on-target editing while limiting off-target effects when used as a ribonucleoprotein complex. Ideal for various CRISPR applications, its proprietary formulation allows for precise genome manipulation, making it a superior choice for research in gene editing.
Catalog No. NC1528809
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.