Learn More
Integrated DNA Technologies Inc Alt-R S.p. HiFi Cas9 Nuclease V3, 100 µg
Supplier:  Integrated DNA Technologies Inc 1081060
Encompass
Recombinant S. pyogenes Cas9 nuclease, purified from an E. coli strain expressing the nuclease. Contains nuclear localization sequence (NLS) and C-terminal 6-His tag. Provided in solution at 10 µg/µL. 100 µg of Cas9 nuclease = 610 pmol.
Catalog No. NC1608092
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.