Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alta Diagnostics Urine Dipstick Controls
SDP

Catalog No. 23770892 Shop All Alta Diagnostics, Inc. Products
Encompass
Change view
Click to view available options
Quantity:
6 x 20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-770892 6 x 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-770892 Supplier Alta Diagnostics Supplier No. 30112
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed specifically to react with commercial dipsticks to register listed responses on the color pads

  • Format: Liquid Semiquantitative Assay
  • Test Ranges: Normal/Abnormal
  • Total number of vials: 6; 3 Normal, 3 Abnormal
  • Volume per vial: 20mL (0.67oz)

Specifications

Content And Storage Room Temperature
Type Urine Dipstick Controls
Intended Use Urinalysis quality controls
Level 3 Normal or 3 Abnormal Level 2
Quantity 6 x 20 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.