Learn More
Medchemexpress LLC Altanserin | 76330-71-7 | 99.9% | 411.5 g/mol | C22H22FN3O2S | 25 MG
SDP
Catalog No. 5000228006
Encompass_Preferred
Altanserin is a selective 5-HT2 receptor ligand used as a research reagent and as a precursor for synthesizing Fluorine-18 labeled altanserin for PET imaging of brain 5-HT2 receptors. It is supplied in high-purity solid and solution formats and is typically prepared in DMSO for experiments.
  • High reported purity: 99.93%.
  • Available as solid quantities (multiple milligram sizes) and as a 10 mM solution in DMSO.
  • Suitable for radiolabeling to produce Fluorine-18 altanserin for PET studies.
  • Soluble in DMSO for straightforward stock preparation.
  • Intended for research use only; not for clinical use.
Catalog No. 50-002-28006 Supplier Medchemexpress LLC Supplier No. HY11915625MG
Altanserin is a selective 5-HT2 receptor ligand used as a research reagent and as a precursor for synthesizing Fluorine-18 labeled altanserin for PET imaging of brain 5-HT2 receptors. It is supplied in high-purity solid and solution formats and is typically prepared in DMSO for experiments.
  • High reported purity: 99.93%.
  • Available as solid quantities (multiple milligram sizes) and as a 10 mM solution in DMSO.
  • Suitable for radiolabeling to produce Fluorine-18 altanserin for PET studies.
  • Soluble in DMSO for straightforward stock preparation.
  • Intended for research use only; not for clinical use.