Learn More
Medchemexpress LLC Altanserin | 76330-71-7 | MFCD00868052 | 99.9% | 411.49 g/mol | C22H22FN3O2S | 5 MG
SDP
Catalog No. 5000228660
Encompass_Preferred
Altanserin is a serotonin 5-HT2 receptor ligand used in research applications, commonly as a precursor to synthesize Fluorine-18-labeled altanserin for PET imaging and receptor binding studies. Supplied as a small-scale solid with high reported purity, it is intended for laboratory research use only and not for human or clinical use.
  • Applicable for radiochemistry and PET tracer synthesis.
  • High purity suitable for analytical and pharmacological studies.
  • Available in small solid pack sizes for experimental work.
  • Soluble in DMSO for solution preparation.
  • Provided with CAS identification to support registration and documentation.
Catalog No. 50-002-28660 Supplier Medchemexpress LLC Supplier No. HY1191565MG
Altanserin is a serotonin 5-HT2 receptor ligand used in research applications, commonly as a precursor to synthesize Fluorine-18-labeled altanserin for PET imaging and receptor binding studies. Supplied as a small-scale solid with high reported purity, it is intended for laboratory research use only and not for human or clinical use.
  • Applicable for radiochemistry and PET tracer synthesis.
  • High purity suitable for analytical and pharmacological studies.
  • Available in small solid pack sizes for experimental work.
  • Soluble in DMSO for solution preparation.
  • Provided with CAS identification to support registration and documentation.