Learn More
Medchemexpress LLC Altanserin | 76330-71-7 | MFCD11045280 | 99.9% | 411.5 g/mol | C22H22FN3O2S | 10 MG
SDP
Catalog No. 5000307621
Encompass_Preferred
Altanserin is a quinazoline-derived research compound that selectively binds the serotonin 5-HT2A receptor and is commonly used in receptor binding studies and to produce Fluorine-18 radioligands for PET imaging. It is supplied as a solid for laboratory research.
  • Binds serotonin 5-HT2A receptors and is suitable for receptor binding assays.
  • Used to synthesize Fluorine-18 altanserin for PET imaging studies.
  • High purity suitable for analytical and research applications.
  • Solid, white to light yellow, and soluble in DMSO for stock preparations.
  • Available in small research quantities for assay development and radioligand synthesis.
Catalog No. 50-003-07621 Supplier Medchemexpress LLC Supplier No. HY11915610MG
Altanserin is a quinazoline-derived research compound that selectively binds the serotonin 5-HT2A receptor and is commonly used in receptor binding studies and to produce Fluorine-18 radioligands for PET imaging. It is supplied as a solid for laboratory research.
  • Binds serotonin 5-HT2A receptors and is suitable for receptor binding assays.
  • Used to synthesize Fluorine-18 altanserin for PET imaging studies.
  • High purity suitable for analytical and research applications.
  • Solid, white to light yellow, and soluble in DMSO for stock preparations.
  • Available in small research quantities for assay development and radioligand synthesis.