Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Altanserin hydrochloride, Tocris Bioscience™

Catalog No. 180950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-095-0 50 mg
18-091-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-095-0 Supplier R&D Systems Supplier No. 1809/50
This item is not returnable. View return policy

5-HT2A receptor antagonist

Potent and selective 5-HT2A receptor antagonist (Ki values are 0.13, 4.55, 40, 62 and 1570 nM at 5-HT2A, α1, 5-HT2C, D2 and 5-HT1A respectively). Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 1135280-78-2
Molecular Formula C22H23ClFN3O2S
Molecular Weight (g/mol) 447.953
InChI Key JFPPLMAMMZZOEA-UHFFFAOYSA-N
Synonym altanserin hydrochloride, unii-xul9717f5k, dsstox_cid_22592, dsstox_rid_80054, dsstox_gsid_42592, 4 1h-quinazolinone, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-, hydrochloride 1:1, 3-2-4-4-fluorobenzoyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorophenyl carbonyl piperidin-1-yl ethyl-2-thioxo-2,3-dihydroquinazolin-4 1h-one hydrochloride, 3-2-4-4-fluorobenzoyl-1-piperidinyl ethyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone hydrochloride
PubChem CID 24978536
IUPAC Name 3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-2-sulfanylidene-1H-quinazolin-4-one;hydrochloride
SMILES C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=S.Cl

Specifications

Quantity 50 mg
Formula Weight 447.95
Percent Purity >98%
Chemical Name or Material Altanserin hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.