Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - Internal thread Matrix/Nunc Bank-It (96 SBS format)
Catalog No. 502719967
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with Matrix/Nunc Bank-It vials - part numbers 3748-BR, 374078, 374074Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9967 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404113
Semi-automated single decapper. Electric capping & decapping. Compatible with Matrix/Nunc Bank-It vials - part numbers 3748-BR, 374078, 374074Includes: Manual, stand, USB charging cable & cap driver.