Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - Sarstedt tubes (custom rack available)
Catalog No. 502719963
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with with Sarstedt tubes (custom rack available)Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9963 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404117
Semi-automated single decapper. Electric capping & decapping. Compatible with with Sarstedt tubes (custom rack available)Includes: Manual, stand, USB charging cable & cap driver.