Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - External thread NEST screw caps (48 SBS rack format)
Catalog No. 502719964
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 48 SBS rack format NEST - external thread screw caps - part numbers 61204x, 61404xIncludes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9964 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404116
Semi-automated single decapper. Electric capping & decapping. Compatible with 48 SBS rack format NEST - external thread screw caps - part numbers 61204x, 61404xIncludes: Manual, stand, USB charging cable & cap driver.