Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - External thread LVL screw caps (24 SBS rack format)
Catalog No. 502719975
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 24 SBS rack format LVL external thread screw caps - part numbers SC24-XT-xxx-xx-xx-x.Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9975 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404105
Semi-automated single decapper. Electric capping & decapping. Compatible with 24 SBS rack format LVL external thread screw caps - part numbers SC24-XT-xxx-xx-xx-x.Includes: Manual, stand, USB charging cable & cap driver.