Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - External thread LVL screw caps (48 SBS rack format)
Catalog No. 502719968
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 48 SBS rack format LVL - external thread screw caps - part numbers SC48-XT-BLUC-NS-CR-LIncludes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9968 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404112
Semi-automated single decapper. Electric capping & decapping. Compatible with 48 SBS rack format LVL - external thread screw caps - part numbers SC48-XT-BLUC-NS-CR-LIncludes: Manual, stand, USB charging cable & cap driver.