Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - Internal thread Matrix (96 SBS format)
Catalog No. 502719979
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with Matrix internal thread screw caps - part number 4470(Colour).Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9979 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404100
Semi-automated single decapper. Electric capping & decapping. Compatible with Matrix internal thread screw caps - part number 4470(Colour).Includes: Manual, stand, USB charging cable & cap driver.