Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - Internal thread Micronic screw caps (24 SBS rack format)
Catalog No. 502719969
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 24 SBS rack format Micronic internal thread screw caps - part numbers MP532XX.Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9969 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404111
Semi-automated single decapper. Electric capping & decapping. Compatible with 24 SBS rack format Micronic internal thread screw caps - part numbers MP532XX.Includes: Manual, stand, USB charging cable & cap driver.