Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - Internal thread fluidX screw caps (96 SBS format)
Catalog No. 502719970
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format fluidX internal thread screw caps - part number 66-63100-Y1 (Y1 to Y13 depending on colour)Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9970 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404110
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format fluidX internal thread screw caps - part number 66-63100-Y1 (Y1 to Y13 depending on colour)Includes: Manual, stand, USB charging cable & cap driver.