Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - Internal thread Hamilton screw caps (96 SBS format)
Catalog No. 502719971
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format Hamilton internal thread screw caps - part numbers 10098363Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9971 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404109
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format Hamilton internal thread screw caps - part numbers 10098363Includes: Manual, stand, USB charging cable & cap driver.