Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - External thread fluidX screw caps (96 SBS format)
Catalog No. 502719972
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format fluidX external thread screw caps - part number 68-53100-Z6N (Z1N to Z13N depending on colour)Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9972 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404108
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format fluidX external thread screw caps - part number 68-53100-Z6N (Z1N to Z13N depending on colour)Includes: Manual, stand, USB charging cable & cap driver.