Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - Internal thread Greiner screw caps (96 SBS format)
Supplier: JDM LAB SOLUTIONS LLC 404107
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format Greiner internal thread screw caps - part numbers 976580, 977580, 978580Includes: Manual, stand, USB charging cable & cap driver.