Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - External thread Nunc screw caps (24 SBS rack format)
Catalog No. 502719960
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 24 SBS rack format Nunc - external thread screw caps - part number 374320Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9960 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404121
Semi-automated single decapper. Electric capping & decapping. Compatible with 24 SBS rack format Nunc - external thread screw caps - part number 374320Includes: Manual, stand, USB charging cable & cap driver.