Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - External thread Eppendorf screw caps (96 SBS format)
Catalog No. 502719966
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with Eppendorf external thread screw caps (96 SBS format) - part number E0030079400Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9966 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404114
Semi-automated single decapper. Electric capping & decapping. Compatible with Eppendorf external thread screw caps (96 SBS format) - part number E0030079400Includes: Manual, stand, USB charging cable & cap driver.