Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - AlteTube
Catalog No. 502720006
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with all screw cap AlteTubes.Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-272-0006 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404001
Add to Cart
Add to Cart
Request a Quote
Semi-automated single decapper. Electric capping & decapping. Compatible with all screw cap AlteTubes.Includes: Manual, stand, USB charging cable & cap driver.