Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch Cassette 12/96 - Internal thread Matrix screw caps
Catalog No. 502719999
Encompass
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Matrix internal thread screw caps - part number 4470(Colour)
Catalog No. 50-271-9999 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404014
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Matrix internal thread screw caps - part number 4470(Colour)