Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch Cassette 12/96 - Internal thread Hamilton screw caps
Catalog No. 502719996
Encompass
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Hamilton internal thread screw caps - part numbers 10098363. For use with AlteCap Switch RT (404999) ONLY.
Catalog No. 50-271-9996 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404017
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Hamilton internal thread screw caps - part numbers 10098363. For use with AlteCap Switch RT (404999) ONLY.