Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch Cassette 12/96 - Internal thread Greiner screw caps
Catalog No. 502719991
Encompass
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Greiner internal thread screw caps - part numbers 976580, 977580, 978580. For use with AlteCap Switch RT (404999) ONLY.
Catalog No. 50-271-9991 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404022
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Greiner internal thread screw caps - part numbers 976580, 977580, 978580. For use with AlteCap Switch RT (404999) ONLY.