Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch Cassette 6/24 - External thread Nalgene screw caps
Catalog No. 502719995
Encompass
AlteCap™ Switch cap driver cassette - 6 channel. Compatible with Nalgene external thread cryogenic vials - part number (Thermo) 12784716. Used with custom AlteCap rack part number 407000 (sold separately)
Catalog No. 50-271-9995 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404018
AlteCap™ Switch cap driver cassette - 6 channel. Compatible with Nalgene external thread cryogenic vials - part number (Thermo) 12784716. Used with custom AlteCap rack part number 407000 (sold separately)