Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch Cassette 6/24 - External thread Nunc screw caps
Catalog No. 502719980
Encompass
AlteCap™ Switch cap driver cassette - 6 channel. Compatible with 24 SBS rack format Nunc - external thread screw caps - part number 374320
Catalog No. 50-271-9980 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404039
AlteCap™ Switch cap driver cassette - 6 channel. Compatible with 24 SBS rack format Nunc - external thread screw caps - part number 374320