Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch Cassette 6/24 - External thread fluidX screw caps
Catalog No. 502720000
Encompass
AlteCap™ Switch cap driver cassette - 6 channel. Compatible with 24 SBS rack format fluidX external thread screw caps - part numbers 66-9401 & 66-9402
Catalog No. 50-272-0000 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404013
AlteCap™ Switch cap driver cassette - 6 channel. Compatible with 24 SBS rack format fluidX external thread screw caps - part numbers 66-9401 & 66-9402