Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch Cassette 8/48 - External thread fluidX screw caps
Catalog No. 502719989
Encompass
AlteCap™ Switch cap driver cassette - 8 channel. Compatible with 48 SBS rack format fluidX external thread screw caps - 2D coded jacketed tubes and older tube type with jacket removed, tricoded tubes in racks 65-9451.
Catalog No. 50-271-9989 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404025
AlteCap™ Switch cap driver cassette - 8 channel. Compatible with 48 SBS rack format fluidX external thread screw caps - 2D coded jacketed tubes and older tube type with jacket removed, tricoded tubes in racks 65-9451.