Learn More
JDM LAB SOLUTIONS LLC AlteCap Switch Decapper
Catalog No. NC3734078
Encompass
Semi-automated handheld decapper. Electronic capping & decapping. Compatible with all 24, 48 & 96 SBS rack format AlteTubes & most other manufacturers 24, 48 & 96 SBS rack format screw cap tubes. Cap driver cassette for customer's tube type purchased separately. (AlteTubes or other manufacturers).
Catalog No. NC3734078 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404000
Add to Cart
Edge
Add to Cart
Request a Quote
Semi-automated handheld decapper. Electronic capping & decapping. Compatible with all 24, 48 & 96 SBS rack format AlteTubes & most other manufacturers 24, 48 & 96 SBS rack format screw cap tubes. Cap driver cassette for customer's tube type purchased separately. (AlteTubes or other manufacturers).