Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Switch RT
Catalog No. 502719956
Encompass
Semi-automated handheld decapper. With reduced torque for specific manufacturers tubes & caps. Electronic capping & decapping. Ergonomically designed dual handle. Cap driver cassette for customer's tube type purchased separately. *For use with Switch cassettes 404017 (Hamilton) & 404022 (Greiner) ONLY.
Catalog No. 50-271-9956 Supplier JDM LAB SOLUTIONS LLC Supplier No. 404999
Add to Cart
Add to Cart
Request a Quote
Semi-automated handheld decapper. With reduced torque for specific manufacturers tubes & caps. Electronic capping & decapping. Ergonomically designed dual handle. Cap driver cassette for customer's tube type purchased separately. *For use with Switch cassettes 404017 (Hamilton) & 404022 (Greiner) ONLY.