Learn More
TARGETMOL CHEMICALS INC ALTENUSIN 5MG
SDP
Catalog No. 502579038
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Altenusin is a natural nonsteroidal fungal metabolite as a novel selective agonist of FXR with an EC50 value of 3.2 ± 0.2 uM. purity: 98%
Catalog No. 50-257-9038 Supplier TARGETMOL CHEMICALS INC Supplier No. T88955MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Altenusin is a natural nonsteroidal fungal metabolite as a novel selective agonist of FXR with an EC50 value of 3.2 ± 0.2 uM. purity: 98%