Learn More
JDM LAB SOLUTIONS LLC AlteRead™Intelligent LF Rack Reader
Catalog No. NC3934362
Encompass
Intelligent AlteRead™ LF rack reader performs all decoding and AI-powered detection internally. No software installation required. Simply plug it in, open your web browser, and start reading. Compatible with 2D barcoded tubes from any manufacturer, it offers flexible data export options CSV, XML, & JSON
Catalog No. NC3934362 Supplier JDM LAB SOLUTIONS LLC Supplier No. 256000
Intelligent AlteRead™ LF rack reader performs all decoding and AI-powered detection internally. No software installation required. Simply plug it in, open your web browser, and start reading. Compatible with 2D barcoded tubes from any manufacturer, it offers flexible data export options CSV, XML, & JSON