Learn More
JDM LAB SOLUTIONS LLC AlteRead™Intelligent LF Rack Reader
Supplier:  JDM LAB SOLUTIONS LLC 256002
Encompass
Intelligent AlteRead™ LF rack reader performs all decoding and AI-powered detection internally. No software installation required. Simply plug it in, open your web browser, and start reading. Compatible with 2D barcoded tubes from any manufacturer, it offers flexible data export options CSV, XML, & JSON
Catalog No. NC3934363