Learn More
JDM LAB SOLUTIONS LLC AlteRead Pendant Single tube 2D & 1D barcode reader
Catalog No. 502720007
Encompass
Single tube 2D & 1D barcode wireless pendant reader. LCD display showing 2D and 1D barcodes instantly and download to any application. Stores up to 2000 decoded barcodes. Compatible with all 2D and 1D tubes. Compatible with Mac OS X, Windows and mobile phone operating systems. Warranty: 1 year
Catalog No. 50-272-0007 Supplier JDM LAB SOLUTIONS LLC Supplier No. 255002
Single tube 2D & 1D barcode wireless pendant reader. LCD display showing 2D and 1D barcodes instantly and download to any application. Stores up to 2000 decoded barcodes. Compatible with all 2D and 1D tubes. Compatible with Mac OS X, Windows and mobile phone operating systems. Warranty: 1 year