Learn More
ATCC Alternaria alternata
Catalog No. 502383335
Encompass
An ampoule containing viable cells (may include spores and mycelia) suspended in cryoprotectant.
Catalog No. 50-238-3335 Supplier ATCC Supplier No. 6663
An ampoule containing viable cells (may include spores and mycelia) suspended in cryoprotectant.