Learn More
AdipoGen Alternarienonic acid (1 mg)
SDP
Supplier:  AdipoGen AGCN20493M001
Encompass_Preferred
Chemical. CAS: 1030376-90-9. Formula: C14H14O6. MW: 278.3. Isolated from Alternaria sp. Moderate inhibitor of selected kinases. Shows no cytotoxic activity in the MTT assay.
Catalog No. 50-205-3677